MilliporeSigma
  • The gene coding for erythrocyte protein band 7.2b (EPB72) is located in band q34.1 of human chromosome 9.
The gene coding for erythrocyte protein band 7.2b (EPB72) is located in band q34.1 of human chromosome 9.
Cytogenetics and cell genetics (1993-01-01)
J A Westberg, B Entler, R Prohaska, J P Schröder
ABSTRACT
Human erythrocyte integral membrane protein band 7 (also termed protein 7.2b or stomatin) is involved in the Na+/K+ permeability of red cells. A cDNA clone coding for this protein was used as a probe to determine the chromosomal localization of the gene (EPB72). Southern blot analysis of somatic cell hybrid DNA panels localized the human EPB72 gene to chromosome band 9q34.1.