MilliporeSigma
  • Ascomycotin A, a new citromycetin analogue produced by Ascomycota sp. Ind19F07 isolated from deep sea sediment.
Ascomycotin A, a new citromycetin analogue produced by Ascomycota sp. Ind19F07 isolated from deep sea sediment.
Natural product research (2014-12-30)
Yong-qi Tian, Xiu-ping Lin, Juan Liu, Kumaravel Kaliyaperumal, Wen Ai, Zhi-ran Ju, Bin Yang, Junfeng Wang, Xian-wen Yang, Yonghong Liu
ABSTRACT
A new citromycetin analogue, ascomycotin A (1), together with eight known compounds, wortmannilactone E (2), orcinol (3), orsellinic acid (4), isosclerone (5), (3R,4S)-( - )-4-hydroxymellein (6), diorcinol (7), chaetocyclinone B (8) and 2,5-dimethoxy-3,6-di(p-methoxypheny1)-1,4-benzoquinone (9), was isolated from the fungal strain Ascomycota sp. Ind19F07, which was isolated from the deep sea sediment of the Indian Ocean. The structures of the compounds were established by spectroscopic data including 1D and 2D NMR and HR-ESI-MS. Compounds (1-9) were evaluated for antibacterial activity.
Sigma-Aldrich
Orcinol, 97%