MilliporeSigma
  • Palladium-catalyzed carbonylative Heck reaction of aryl bromides with vinyl ethers to 3-alkoxy alkenones and pyrazoles.
Palladium-catalyzed carbonylative Heck reaction of aryl bromides with vinyl ethers to 3-alkoxy alkenones and pyrazoles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-03-17)
Johannes Schranck, Xiao-Feng Wu, Helfried Neumann, Matthias Beller
ABSTRACT
Three COming together: The first carbonylative Heck coupling reaction of aryl bromides and vinyl ethers leading to 1-aryl-3-alkoxy-2-propen-1-ones has been established (see scheme). Based on this coupling methodology, a novel one-pot synthesis of aryl-substituted pyrazoles was also realized.
Sigma-Aldrich
Palladium(II) bromide, 99%