MilliporeSigma
  • BEGAIN (brain-enriched guanylate kinase-associated protein), a novel neuronal PSD-95/SAP90-binding protein.
BEGAIN (brain-enriched guanylate kinase-associated protein), a novel neuronal PSD-95/SAP90-binding protein.
The Journal of biological chemistry (1998-10-03)
M Deguchi, Y Hata, M Takeuchi, N Ide, K Hirao, I Yao, M Irie, A Toyoda, Y Takai
ABSTRACT
PSD-95/SAP90 is a synaptic membrane-associated guanylate kinase with three PDZ, one SH3, and one guanylate kinase (GK) domain. PSD-95/SAP90 binds various proteins through the PDZ domains and organizes synaptic junctions. PSD-95/SAP90 also interacts with the postsynaptic density (PSD) fraction-enriched protein, named SAPAP (also called GKAP and DAP), through the GK domain. SAPAP is Triton X-100-insoluble and recruits PSD-95/SAP90 into the Triton X-100-insoluble fraction in the transfected cells, suggesting that SAPAP may fix PSD-95/SAP90 to the PSD. Here we report a novel protein interacting with the GK domain of PSD-95/SAP90, BEGAIN. BEGAIN is specifically expressed in brain and enriched in the PSD fraction. BEGAIN is Triton X-100-soluble in the transfected cells but is recruited to the Triton X-100-insoluble fraction by SAPAP when coexpressed with PSD-95/SAP90. BEGAIN may be a novel PSD component associated with the core complex of PSD-95/SAP90 and SAPAP.