MilliporeSigma
  • Interleukin 12 induces tyrosine phosphorylation and activation of STAT4 in human lymphocytes.
Interleukin 12 induces tyrosine phosphorylation and activation of STAT4 in human lymphocytes.
Proceedings of the National Academy of Sciences of the United States of America (1995-08-01)
C M Bacon, E F Petricoin, J R Ortaldo, R C Rees, A C Larner, J A Johnston, J J O'Shea
ABSTRACT
Interleukin 12 (IL-12) is an important immunoregulatory cytokine whose receptor is a member of the hematopoietin receptor superfamily. We have recently demonstrated that stimulation of human T and natural killer cells with IL-12 induces tyrosine phosphorylation of the Janus family tyrosine kinase JAK2 and Tyk2, implicating these kinases in the immediate biochemical response to IL-12. Recently, transcription factors known as STATs (signal transducers and activators of transcription) have been shown to be tyrosine phosphorylated and activated in response to a number of cytokines that bind hematopoietin receptors and activate JAK kinases. In this report we demonstrate that IL-12 induces tyrosine phosphorylation of a recently identified STAT family member, STAT4, and show that STAT4 expression is regulated by T-cell activation. Furthermore, we show that IL-12 stimulates formation of a DNA-binding complex that recognizes a DNA sequence previously shown to bind STAT proteins and that this complex contains STAT4. These data, and the recent demonstration of JAK phosphorylation by IL-12, identify a rapid signal-transduction pathway likely to mediate IL-12-induced gene expression.
Sigma-Aldrich
Cell-Based ELISA for detecting phospho-STAT4 (pTyr693) in cultured cell lines, 96 assays