MilliporeSigma
  • Effect of F-spondin on cementoblastic differentiation of human periodontal ligament cells.
Effect of F-spondin on cementoblastic differentiation of human periodontal ligament cells.
Biochemical and biophysical research communications (2006-09-13)
Masae Kitagawa, Yasusei Kudo, Shinji Iizuka, Ikuko Ogawa, Yoshimitsu Abiko, Mutsumi Miyauchi, Takashi Takata
ABSTRACT
Cementum is a mineralized tissue produced by cementoblasts covering the roots of teeth that provides for the attachment of periodontal ligament to roots and surrounding alveolar bone. To study the mechanism of proliferation and differentiation of cementoblasts is important for understanding periodontal physiology and pathology including periodontal tissue regeneration. However, the detailed mechanism of the proliferation and differentiation of human cementoblasts is still unclear. We previously established human cementoblast-like (HCEM) cell lines. We thought that comparing the transcriptional profiles of HCEM cells and human periodontal ligament (HPL) cells derived from the same teeth could be a good approach to identify genes that influence the nature of cementoblasts. We identified F-spondin as the gene demonstrating the high fold change expression in HCEM cells. Interestingly, F-spondin highly expressing HPL cells showed similar phenotype of cementoblasts, such as up-regulation of mineralized-related genes. Overall, we identified F-spondin as a promoting factor for cementoblastic differentiation.