MilliporeSigma
  • Comparative thermodynamic properties, vibrational spectral studies, NBO and HOMO-LUMO analyses of 3-chloro-2,4,5,6-tetrafluoropyridine and 4-bromo-2,3,5,6-tetrafluoropyridine based on density functional theory.
Comparative thermodynamic properties, vibrational spectral studies, NBO and HOMO-LUMO analyses of 3-chloro-2,4,5,6-tetrafluoropyridine and 4-bromo-2,3,5,6-tetrafluoropyridine based on density functional theory.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2014-05-27)
C Selvarani, V Balachandran, K Vishwanathan
ABSTRACT
Quantum mechanical calculations of energies, geometries and vibrational wave numbers of 3-chloro-2,4,5,6-tetrafluoropyridine and 4-bromo-2,3,5,6-tetrafluoropyridine have been performed by DFT level of theory using B3LYP/6-31+G(d) and B3LYP/6-311++G(d,p) as basis sets. The optimized geometrical parameters obtained by B3LYP method show good agreement with experimental data. The difference between the observed and scaled wave number values of most of the fundamentals is very small. A detailed interpretation of the FT-IR and FT-Raman spectra of 3-chloro-2,4,5,6-tetrafluoropyridine and 4-bromo-2,3,5,6-tetrafluoropyridine were also reported. Molecular stability and bond strength were investigated by applying the natural bond orbital analysis (NBO). The calculated HOMO and LUMO energies show that charge transfer occurs in the molecules. Information about the size, shape, charge density distribution, and site of chemical reactivity of the molecules has been obtained by mapping electron density isosurface with electrostatic potential (ESP). Thermodynamic properties (heat capacity, entropy and enthalpy and Gibb's free energy) of the title compounds at different temperatures were calculated.
Sigma-Aldrich
4-Bromo-2,3,5,6-tetrafluoropyridine, 98%