MilliporeSigma
  • Antimicrobial activity of some thiourea derivatives and their nickel and copper complexes.
Antimicrobial activity of some thiourea derivatives and their nickel and copper complexes.
Molecules (Basel, Switzerland) (2009-01-27)
Hakan Arslan, Nizami Duran, Gulay Borekci, Cemal Koray Ozer, Cevdet Akbay
ABSTRACT
Five thiourea derivative ligands and their Ni(2+) and Cu(2+) complexes have been synthesized. The compounds were screened for their in vitro anti-bacterial activity using Gram-positive bacteria (two different standard strains of Staphylococcus aureus, Staphylococcus epidermidis, Enterococcus faecalis, Streptococcus pyogenes, Bacillus cereus) and Gram-negative bacteria (Esherichia coli, Pseudomonas aeruginosa, Enterobacter cloacae, Proteus vulgaris, Enterobacter aerogenes) and in vitro anti-yeast activity (Candida albicans, Candida krusei, Candida glabrata, Candida tropicalis, Candida parapsilosis). The minimum inhibitory concentration was determined for all ligands and their complexes. In vitro anti-yeast activity of both ligands and their metal complexes is greater than their in vitro anti-bacterial activity. The effect of the structure of the investigated compounds on the antimicrobial activity is discussed.
Sigma-Aldrich
Cyclohexanecarbonyl chloride, 98%