MilliporeSigma
[Clinical studies with levodopa/carbidopa intestinal gel].
Ideggyogyaszati szemle (2014-03-25)
Péter Klivényi, László Vécsei
ABSTRACT
The motor complications in advanced Parkinson's disease may interfere with the quality of life. To overcome this disability, a jejunal levodopa pump was developed. In this review we have summarized the published clinical data with this method.
Product Description
Carbidopa, European Pharmacopoeia (EP) Reference Standard
USP
Carbidopa, United States Pharmacopeia (USP) Reference Standard