MilliporeSigma
Capillary electrophoresis.
Science (New York, N.Y.) (1988-10-14)
M J Gordon, X Huang, S L Pentoney, R N Zare
ABSTRACT
Electrophoretic separations carried out in capillary tubes offer the possibilities of rapid and automated analyses of small volumes of complex mixtures with unprecedented resolution and sensitivity. Some emerging developments of this new instrumentation are reviewed.