MilliporeSigma
  • Disturbances of Wnt/β-catenin pathway and energy metabolism in early CKD: effect of phosphate binders.
Disturbances of Wnt/β-catenin pathway and energy metabolism in early CKD: effect of phosphate binders.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2013-08-27)
Rodrigo B de Oliveira, Fabiana G Graciolli, Luciene M dos Reis, Ana L E Cancela, Lilian Cuppari, Maria E Canziani, Aluizio B Carvalho, Vanda Jorgetti, Rosa M A Moysés
ABSTRACT
Mineral bone disorder (MBD) is an early complication of chronic kidney disease (CKD), with complex interactions in the bone-kidney-energy axis. These events lead to impaired bone remodelling, which in turn is associated with cardiovascular disease. Recently, we reported on a positive effect of phosphate binder treatment on bone remodelling markers and a reduction in serum FGF-23 levels in predialysis-CKD patients. The goal of the present study of this trial was to examine the effects of phosphate binders on energy-regulating hormones and Wnt pathway. In this present post hoc analysis of the above randomized, open-label, 8-week trial, which compared the effects of increasing doses of sevelamer-HCl or calcium acetate on various CKD-MBD parameters in 40 normophosphatemic CKD Stage 3-4 patients, we measured serum sclerostin, Dickkopf-1, leptin, adiponectin and serotonin concentrations. Serum sclerostin, Dickkopf-1 and leptin were elevated at baseline despite normal calcium, phosphorus levels and daily urinary phosphorus excretion. There were significant and positive correlations between sclerostin and FGF-23, as well between leptin and Dickkopf-1. Treatment with both phosphate binders led to a significant decrease in phosphate overload. However, sevelamer-HCl, but not with calcium acetate, led to a significant decrease in serum FGF-23, sclerostin and leptin, and to a significant increase in bone alkaline phosphatase levels. Early stages of CKD are associated with an impairment of the Wnt pathway, as reflected by elevated sclerostin, and a dysregulation of energy-regulating hormones. Many of these disturbances can be ameliorated by phosphate binder treatment, more with sevelamer-HCl than with calcium acetate.
Product Description
Supelco
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material