MilliporeSigma
  • Concomitant endoureterotomy and dextranomer/hyaluronic acid subureteral injection for management of obstructive refluxing megaureter.
Concomitant endoureterotomy and dextranomer/hyaluronic acid subureteral injection for management of obstructive refluxing megaureter.
Journal of endourology (2011-11-09)
Abdol-Mohammad Kajbafzadeh, Ali Tourchi
ABSTRACT
To present the results of our experience with combined endoureterotomy and endoscopic injection of dextranomer/hyaluronic acid (Deflux) for the treatment of primary obstructive refluxing megaureter (PORM). Eighteen children (12 female, 6 male; mean age-14 months) with 20 PORM units underwent concomitant endoureterotomy and endoscopic subureteral Deflux injection. All patients underwent endoureterotomy at the 6-o'clock position with insertion of a 3F Double-J ureteral stent into the obstructed segment of ureter and subureteral injection of Deflux at the 5-o'clock and 7-o'clock positions. The Double-J stent was left in place with its distal tip fixed with a single knot to the external genitalia for easy removal after 1 week. Patients with refluxing nonobstructive ureter on the contralateral side of the PORM unit (seven children) underwent simultaneous endoscopic subureteral injection of Deflux. Voiding cystourethrography (VCUG) was performed at 6 months, and ultrasonography was performed at 1 week 3, 6, and 12 months postoperatively. With a mean follow-up of 30 months, the procedure was uneventful in all patients. Follow-up VCUG showed no evidence of reflux in 15 ureterorenal (75%), significant decrease in reflux grade in 2 (10%), and no change in 3 (15%) in the endoscopic treated PORM units. No evidence of reflux was observed in the treated contralateral refluxing nonobstructive ureters. Ultrasonography revealed no ureterovesical junction obstruction. In 19 ureterorenal (95%) units, there was a complete resolution or decrease in hydroureteronephrosis. The results of this study demonstrate that combined endoureterotomy and subureteral injection of Deflux is safe and effective in the treatment of PORM in selected patients.
Product Description
Dextranomer, European Pharmacopoeia (EP) Reference Standard