MilliporeSigma
  • Insecticidal activity of chemical constituents from Aristolochia pubescens against Anticarsia gemmatalis larvae.
Insecticidal activity of chemical constituents from Aristolochia pubescens against Anticarsia gemmatalis larvae.
Pest management science (2004-05-04)
Isabele R Nascimento, Afonso T Murata, Sergio A Bortoli, Lucia M X Lopes
ABSTRACT
Acetone and ethanol extracts of the tubercula and several compounds isolated from Aristolochia pubescens (Willd) were bioassayed on velvetbean caterpillars, Anticarsia gemmatalis (Hübner), for evaluation of the insecticidal activities. Of the extracts subjected to bioassay, the acetone extract showed the highest activity. (-)-Cubebin did not show activity against soybean caterpillars, whereas aristolochic acid and ent-kaur-15-en-17-ol increased the larval period. These compounds, and (+)-eudesmin and (+)-sesamin, reduced the viability of this period, giving rise to malformed adults. These extracts and compounds are therefore potential botanical insecticide agents for the control of velvetbean caterpillars in soybean crops.
Sigma-Aldrich
Eudesmine, ≥95% (LC/MS-ELSD)