MilliporeSigma
[Determination of nitidine in different parts of Zanthoxylum nitidum].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2002-01-22)
S Zhang, Y Yao, C Liu
ABSTRACT
To determine the nitidine in different parts of Zanthoxylum nitidum (Roxb.) DC. HPLC method was used. The column was HYPERSIL BDS C18. The mobile phase was 0.02 mol/L monobasic potassium phosphate: acetonitrile (66:34). The detective wavelength was 328 nm. The average recoveries and RSD for nitidine was 100.24%, 1.83% respectively. The root contained more nitidine than other parts. The method can be used for the quality control of Zanthoxylum nitidum (Roxb.) DC. The roots were the best part for medicine.