MilliporeSigma
  • Alpha-dihydroergocryptine and predictive factors in migraine prophylaxis.
Alpha-dihydroergocryptine and predictive factors in migraine prophylaxis.
International journal of clinical pharmacology and therapeutics (2001-05-03)
G Micieli, A Cavallini, S Marcheselli, F Mailland, L Ambrosoli, G Nappi
ABSTRACT
A double-blind, crossover study was carried out to compare the efficacy of alpha-dihydroergocryptine mesylate (10 mg twice daily) vs propranolol (40 mg twice daily) in the prophylaxis of migraine without aura, and to identify possible predictors of therapeutic response by evaluating the symptomatological profile of individual migraine attacks and the autonomic cardiovascular response to noradrenergic and dopaminergic (cold pressor, bromocriptine) tests. Forty migraineurs (10 males, 30 females) were randomized according to a two-period (3-month), two-treatment, crossover design. Efficacy was assessed using quantitative data recorded in the patient's headache diary. Data were evaluated using the Wallenstein's method. Both drugs showed a significant reduction in all the efficacy variables (headache attacks, days with headache, analgesic consumption) with no difference between treatments. Neither a bromocriptine test, nor a cold pressor test nor the symptomatological profile of individual migraine attacks differed between the two groups of migraine patients. Ten patients experienced at least one adverse drug reaction during the first period of the crossover design, 5 being treated with alpha-dihydroergocryptine and 5 with propranolol. It is concluded that alpha-dihydroergocryptine is an effective medication for migraine prophylaxis. The biochemical tests and the type of psychological profile cannot be used to predict drug response.
Product Description
Codergocrine mesilate, European Pharmacopoeia (EP) Reference Standard