MilliporeSigma
  • [2-Chloro-1-methylpyridinium iodide. A suitable reagent for peptide synthesis].
[2-Chloro-1-methylpyridinium iodide. A suitable reagent for peptide synthesis].
Biological chemistry Hoppe-Seyler (1985-12-01)
W Keese, H Khalaf, D Grambow, G Grundke, M Rimpler
ABSTRACT
2-Chloro-1-methylpyridinium iodide is a coupling reagent for peptide synthesis. By its use different peptides were synthesized with protected di- and trifunctional amino acids. While urethan-protected amino acids react free of racemization, the fragment condensation needs N-Hydroxysuccinimide, as shown by the Young test.
Sigma-Aldrich
2-Chloro-1-methylpyridinium iodide, 97%