MilliporeSigma
  • Mass transfer and adsorption equilibrium for low volatility alkanes in BPL activated carbon.
Mass transfer and adsorption equilibrium for low volatility alkanes in BPL activated carbon.
Langmuir : the ACS journal of surfaces and colloids (2013-01-31)
Yu Wang, John J Mahle, Amanda M B Furtado, T Grant Glover, James H Buchanan, Gregory W Peterson, M Douglas LeVan
ABSTRACT
The structure of a molecule and its concentration can strongly influence diffusional properties for transport in nanoporous materials. We study mass transfer of alkanes in BPL activated carbon using the concentration-swing frequency response method, which can easily discriminate among mass transfer mechanisms. We measure concentration-dependent diffusion rates for n-hexane, n-octane, n-decane, 2,7-dimethyloctane, and cyclodecane, which have different carbon numbers and geometries: straight chain, branched chain, and cyclic. Micropore diffusion is determined to be the controlling mass transfer resistance except at low relative saturation for n-decane, where an external mass transfer resistance also becomes important, showing that the controlling mass transfer mechanism can change with system concentration. Micropore diffusion coefficients are found to be strongly concentration dependent. Adsorption isotherm slopes obtained from measured isotherms, the concentration-swing frequency response method, and a predictive method show reasonably good agreement.