MilliporeSigma
  • Non-imidazole histamine H3 ligands, part IV: SAR of 1-[2-thiazol-5-yl-(2-aminoethyl)]-4-n-propylpiperazine derivatives.
Non-imidazole histamine H3 ligands, part IV: SAR of 1-[2-thiazol-5-yl-(2-aminoethyl)]-4-n-propylpiperazine derivatives.
European journal of medicinal chemistry (2008-11-08)
Anna Frymarkiewicz, Krzysztof Walczyński
ABSTRACT
A series of 1-[[2-thiazol-5-yl-(2-aminoethyl)]-4-n-propyl]piperazine derivatives have been prepared and in vitro tested as H(3)-receptor antagonists (the electrically evoked contraction of the guinea pig jejunum). It appeared that by comparison of homologous pairs the 1-[[2-thiazol-5-yl-(2-methyl-2-phenylalkylaminoethyl)]-4-n-propyl]piperazine derivatives (4c1-4c3) have slightly higher activity than their 1-[2-thiazol-5-yl-(2-methyl-2-alkylaminoethyl)]-4-n-propylpiperazine analogues (4b1-4b3). In the 2-methylalkylamide series (4a1-4a3) a somewhat lower activity was observed. The most potent compound of the series is the 1-[2-thiazol-5-yl-(2-methyl-2-phenylpropylaminoethyl)]-4-n-propylpiperazine (4c2) with pA(2)=8.27 (its alkyl analogue (4b2) showed pA(2)=7.53 and the corresponding amide (4a2) displayed pA(2)=7.36). Selected compounds (4b1, 4b2, 4c1 and 4c2) were also tested (in vitro) for H(1) antagonistic effects in vitro applying standard methods (guinea pig ileum). None showed any H(1) antagonistic activity (pA(2)<4).
Sigma-Aldrich
1-n-Propylpiperazine dihydrobromide, 96%