MilliporeSigma
  • Quantification of busulfan in plasma by gas chromatography-mass spectrometry following derivatization with tetrafluorothiophenol.
Quantification of busulfan in plasma by gas chromatography-mass spectrometry following derivatization with tetrafluorothiophenol.
Journal of chromatography. B, Biomedical sciences and applications (1998-07-08)
M H Quernin, B Poonkuzhali, C Montes, R Krishnamoorthy, D Dennison, A Srivastava, E Vilmer, M Chandy, E Jacqz-Aigrain
ABSTRACT
A specific and highly sensitive method has been developed for the determination of busulfan in plasma by gas chromatography-mass spectrometry using a deuterium-labeled busulfan (busulfan-d8) as internal standard. Plasma containing busulfan and busulfan-d8 were extracted with ethyl acetate and derivatized with 2,3,5,6-tetrafluorothiophenol prior to the monitoring of specific ions. The limit of quantification of the assay was 20 ng/ml and the calibration curve was linear over the range of 10 to 2000 ng/ml of derivatized busulfan. This method was in good agreement with the GC-MS assay using derivatization with sodium iodide and measuring diiodobutane. In addition, a pharmacokinetic study of busulfan was conducted in six children. The apparent oral clearance was 5.7+/-1.9 ml/kg/min and the volume of distribution was 1.0+/-0.4 l/kg and were similar to those previously reported in pediatric patients.
Sigma-Aldrich
2,3,5,6-Tetrafluorobenzenethiol, 97%