MilliporeSigma
  • Accumulation of indoxyl-beta-D-glucuronide in uremic serum: suppression of its production by oral sorbent and efficient removal by hemodialysis.
Accumulation of indoxyl-beta-D-glucuronide in uremic serum: suppression of its production by oral sorbent and efficient removal by hemodialysis.
Nephron (1996-01-01)
T Niwa, T Miyazaki, S Tsukushi, K Maeda, Y Tsubakihara, A Owada, T Shiigai
ABSTRACT
We identified and quantified indoxyl-beta-D-glucuronide in uremic serum and urine to determine the metabolism of indoles including indoxyl sulfate in uremic patients. Serum levels of indoxyl-beta-D-glucuronide were markedly increased in undialyzed uremic patients, in patients on hemodialysis, and in patients on continuous ambulatory peritoneal dialysis. Urinary excretion of indoxyl-beta-D-glucuronide was also increased in undialyzed uremic patients. Urinary indoxyl-beta-D-glucuronide was significantly correlated with serum indoxyl sulfate, indicating that a high serum level of indoxyl sulfate leads to the enhanced synthesis of indoxyl-beta-D-glucuronide. Oral sorbent (AST-120) administration markedly decreased the serum and urine levels of indoxyl-beta-D-glucuronide as well as indoxyl sulfate in the undialyzed uremic patients. Serum indoxyl-beta-D-glucuronide could be efficiently removed by hemodialysis despite its high protein-binding ratio of about 50%. In conclusion, the serum level of indoxyl-beta-D-glucuronide increases in uremic patients due to renal insufficiency and its increased production. The production of indoxyl-beta-D-glucuronide can be suppressed by oral sorbent treatment, and serum indoxyl-beta-D-glucuronide can be efficiently removed by hemodialysis.
Sigma-Aldrich
Indoxyl β-D-glucuronide cyclohexylammonium salt