MilliporeSigma
  • Reduction of 1-nitroso-2-naphthol by NADPH in the presence of liver microsomes.
Reduction of 1-nitroso-2-naphthol by NADPH in the presence of liver microsomes.
The International journal of biochemistry (1993-02-01)
V Leskovac, J O Peggins, S Trivić, J Svircević, M Popović, M Stupar
ABSTRACT
1. The endogenous, NADPH-supported production of H2O2 and of O2-.-radicals in liver microsomes, was very strongly enhanced in the presence of 1-nitroso-2-naphthol. 2. A 30-fold induction by NON was the consequence of its direct reduction to NON-radicals, catalyzed by microsomal NADPH:cytochrome P450 reductase. 3. Nitroso radicals reduce molecular oxygen to superoxide anion radicals, which were readily dismutated by superoxide dismutase to hydrogen peroxide. 4. O2-.-radicals were the sole precursors of all NON-induced production of H2O2 in liver microsomes.
Sigma-Aldrich
1-Nitroso-2-naphthol, 97%