MilliporeSigma
  • Crystallization and preliminary crystallographic analysis of a DNA (cytosine-5)-methyltransferase from Haemophilus aegyptius bound covalently to DNA.
Crystallization and preliminary crystallographic analysis of a DNA (cytosine-5)-methyltransferase from Haemophilus aegyptius bound covalently to DNA.
Journal of molecular biology (1994-05-13)
K M Reinisch, L Chen, G L Verdine, W N Lipscomb
ABSTRACT
A DNA (cytosine)-5-methyltransferase from Haemophilus aegyptius (M.Hae III), which catalyzes methyl transfer from S-adenosyl-L-methionine to DNA, has been crystallized as a covalent complex with a suicide oligonucleotide substrate. Crystals of the co-complex were grown by vapor diffusion with hanging droplets, using polyethylene glycol 3500 as the precipitant. The crystals belong to the orthorhombic space group P2(1)2(1)2(1); the unit cell parameters are a = 57.6 A, b = 108.0 A, c = 155.8 A with two protein-DNA complexes in the asymmetric unit. Complete sets of native and derivative data have been collected to 2.7 A using a laboratory source.
Sigma-Aldrich
5−Fluoro−2′−deoxycytidine, ≥98% (HPLC), powder