MilliporeSigma
  • Dynamic differentiation of GABAA-sensitive influences on orientation selectivity of complex cells in the cat striate cortex.
Dynamic differentiation of GABAA-sensitive influences on orientation selectivity of complex cells in the cat striate cortex.
Experimental brain research (1995-01-01)
B Pfleger, A B Bonds
ABSTRACT
The influence of GABAA receptors on orientation selectivity of cat complex cells was tested by iontophoresis of the GABAA receptor blockers bicuculline and N-methyl-bicuculline while stimulating with drifting sinusoidal gratings. Reduction of orientation tuning was markedly less than reported in previous studies that used drifting bars as visual stimuli. Only 3/31 cells lost orientation selectivity, with an average increase in bandwidth of 33%, as opposed to half the cells losing selectivity and a bandwidth increase for the remainder of 47% as reported previously. Infusion of GABAA blockers revealed a prominent stimulus onset transient response, lasting about 120 ms, that showed a broadening of orientation selectivity comparable to that found using drifting bars under similar circumstances. We believe that drifting gratings emphasize a steady-state response component that retains, in the presence of GABAA blockers, significant orientation selectivity. Because the onset transient is initially unselective for orientation, we suggest that the steady-state, orientation-selective response component develops from an alternate inhibitory mechanism, possibly mediated by GABAB receptors.
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methchloride, ≥97% (HPLC), powder