MilliporeSigma
  • Liquid chromatographic method for determination of citreoviridin in corn and rice.
Liquid chromatographic method for determination of citreoviridin in corn and rice.
Journal - Association of Official Analytical Chemists (1988-07-01)
R D Stubblefield, J I Greer, O L Shotwell
ABSTRACT
Citreoviridin, a neurotoxic mycotoxin, has been found as a natural contaminant in corn left unharvested in the southeastern United States and in rice of several Asian countries, including Japan. A reliable analytical method for the quantitative determination of citreoviridin in corn and rice is described. Corn or rice is extracted with dichloromethane, and the extract is partially purified on silica and amino solid-phase extraction (SPE) columns. The extract is analyzed for citreoviridin by normal-phase liquid chromatography, using a mobile phase of ethyl acetate-hexane (75 + 25) at 1.5 mL/min and a fluorescence detector to measure the yellow fluorescence (388 nm excitation, 480 nm emission). With a 100 microL injection loop, the relationship between concentration and injection volume is linear for 20-60 microL injections. Recoveries of citreoviridin added to yellow corn at 10-50 ng/g were 91.0-96.9%; recoveries from white corn (10-50 ng/g added) were 96.8-102.8%. Recoveries of 5000 ng/g added to white corn were 89.0%, indicating that heavily contaminated samples can be assayed by the method. Minimum detection limits were 10 ng for citreoviridin standard and 2 ng/g for citreoviridin added to corn. White rice fermented with Penicillium citreo-viride (1524 ppm) was mixed with and serially diluted with uncontaminated ground corn to obtain citreoviridin-contaminated corn (ca 25 ppb). When the samples were assayed by the method, a mean level of 24.4 +/- 1.65 ppb (6.5% coefficient of variation) was obtained. Four fermented rice food samples and 3 commercial rice samples were investigated.(ABSTRACT TRUNCATED AT 250 WORDS)