MilliporeSigma
  • Fungitoxicity of m-fluorophenylalanine-containing peptides towards Pythium ultimum.
Fungitoxicity of m-fluorophenylalanine-containing peptides towards Pythium ultimum.
Experientia (1989-04-15)
D H Young, R J Mehta
ABSTRACT
The tripeptide L-m-fluorophenylalanyl-L-alanyl-L-alanine was much more fungitoxic towards Pythium ultimum than the dipeptide L-m-fluorophenylalanyl-L-alanine or m-fluorophenylalanine. The fungitoxicity of the tripeptide was reduced by L-alanyl peptides and phenylalanine, but not by other amino acids. In contrast, the fungitoxicity of m-fluorophenylalanine was unaffected by peptides, and was antagonized by several amino acids. These results suggest the effective delivery of m-fluorophenylalanine into the cell by a tripeptide carrier.
Sigma-Aldrich
m-Fluoro-DL-phenylalanine