MilliporeSigma
  • 2,3,7,8-Tetrachlorodibenzo-p-dioxin promotes BHV-1 infection in mammalian cells by interfering with iron homeostasis regulation.
2,3,7,8-Tetrachlorodibenzo-p-dioxin promotes BHV-1 infection in mammalian cells by interfering with iron homeostasis regulation.
PloS one (2013-03-23)
Filomena Fiorito, Carlo Irace, Antonio Di Pascale, Alfredo Colonna, Giuseppe Iovane, Ugo Pagnini, Rita Santamaria, Luisa De Martino
ABSTRACT
Mammalian cells require iron to satisfy metabolic needs or to accomplish specialized functions, and DNA viruses, like bovine herpesvirus 1 (BHV-1), require an iron-replete host to efficiently replicate, so that iron bioavailability is an important component of viral virulence. Cellular iron metabolism is coordinately controlled by the Iron Regulatory Proteins (IRP1 and IRP2), whose activity is affected by 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD), a current and persistent environmental contaminant. Considering that TCDD enhances BHV-1 replication, herein we analyzed the effects of TCDD on iron metabolism during BHV-1 infection in MDBK cells, and presented evidences of a divergent modulation of IRP1 and IRP2 RNA-binding capacity. Moreover, an up-regulation of transferrin receptor 1 (TfR1) and a concomitant down-regulation of ferritin were observed. This scenario led to an expansion of the labile iron pool (LIP) and induces a significant enhance of viral titer, as confirmed by increased levels of BHV-1 infected cell protein 0 (bICP0), the major transcriptional regulatory protein of BHV-1. Taken together, our data suggest that TCDD increases the free intracellular iron availability thereby promoting the onset of BHV-1 infection and rendering bovine cells more vulnerable to the virus.
Sigma-Aldrich
Aconitase from porcine heart
Sigma-Aldrich
MISSION® esiRNA, targeting human ACO1
Sigma-Aldrich
MISSION® esiRNA, targeting human TFRC
Sigma-Aldrich
MISSION® esiRNA, targeting human IREB2