MilliporeSigma
  • Phase III study of ACVBP versus ACVBP plus rituximab for patients with localized low-risk diffuse large B-cell lymphoma (LNH03-1B).
Phase III study of ACVBP versus ACVBP plus rituximab for patients with localized low-risk diffuse large B-cell lymphoma (LNH03-1B).
Annals of oncology : official journal of the European Society for Medical Oncology (2012-12-14)
N Ketterer, B Coiffier, C Thieblemont, C Fermé, J Brière, O Casasnovas, S Bologna, B Christian, T Connerotte, C Récher, D Bordessoule, C Fruchart, R Delarue, C Bonnet, F Morschhauser, B Anglaret, C Soussain, B Fabiani, H Tilly, C Haioun
ABSTRACT
The superiority of a chemotherapy with doxorubicin, cyclophosphamide, vindesine, bleomycin and prednisone (ACVBP) in comparison with cyclophosphamide, doxorubicin, vincristin and prednisone plus radiotherapy for young patients with localized diffuse large B-cell lymphoma (DLBCL) was previously demonstrated. We report the results of a trial which evaluates the role of rituximab combined with ACVBP (R-ACVBP) in these patients. Untreated patients younger than 66 years with stage I or II DLBCL and no adverse prognostic factors of the age-adjusted International Prognostic Index were randomly assigned to receive three cycles of ACVBP plus sequential consolidation with or without the addition of four infusions of rituximab. A total of 223 patients were randomly allocated to the study, 110 in the R-ACVBP group and 113 in the ACVBP group. After a median follow-up of 43 months, our 3-year estimate of event-free survival was 93% in the R-ACVBP group and 82% in the ACVBP group (P = 0.0487). Three-year estimate of progression-free survival was increased in the R-ACVBP group (95% versus 83%, P = 0.0205). Overall survival did not differ between the two groups with a 3-year estimates of 98% and 97%, respectively (P = 0.686). In young patients with low-risk localized DLBCL, rituximab combined with three cycles of ACVBP plus consolidation is significantly superior to ACVBP plus consolidation alone.
Sigma-Aldrich
Vindesine sulfate salt hydrate, ≥95% (HPLC)