- A rare cause of chronic rectal bleeding in children; solitary rectal ulcer: case report.
A rare cause of chronic rectal bleeding in children; solitary rectal ulcer: case report.
Solitary rectal ulcer causing lower gastrointestinal bleeding is extremely rare in children. Rare presentation, non-specific symptoms, insufficient experience, and characteristics mimicking other rectal diseases may cause misdiagnosis or delay of diagnosis in some pediatric patients. Here, we report a 10-year-old boy with solitary rectal ulcer diagnosed two years after onset of the symptoms who responded well to the conservative therapy, including high-fiber diet, laxatives, defecation training, and sucralfate enema.