MilliporeSigma
  • Dentin-bonding molar efficiency using N-phenylglycine, N-phenyl-beta-alanine, or N-methyl-N-phenylglycine.
Dentin-bonding molar efficiency using N-phenylglycine, N-phenyl-beta-alanine, or N-methyl-N-phenylglycine.
Journal of dental research (1991-03-01)
R E Webb, A D Johnston
ABSTRACT
Three structurally related substituted amino acids (N-compounds) were studied in a three-step dentin-bonding protocol. The first step of an acidic ferric oxalate solution and the third step of a surface-active comonomer were held constant throughout the study. In the second step, the amount of the N-compound--either N-phenylglycine (NPG), N-methyl-N-phenylglycine (NMNPG), or N-phenyl-beta-alanine (NPBA)--was varied in acetone from 0 mol/L through 5 x 10(-1) mol/L in seven steps. At 1 x 10(-3) mol/L for NPG and NMNPG, average bond strength values were 7.4 +/- 2.2 and 10.5 +/- 2.7 MPa. The highest bond strength value for NMNPG was at 1 x 10(-2) mol/L, with 13.2 +/- 4.0 MPa. The highest value for NPG was at 1 x 10(-1) mol/L, with a value of 11.8 +/- 2.5 MPa. The average bond strength for NPBA did not differ from zero across the entire range of concentrations. Molar efficiency was defined as the bond strength per mole of these applied N-compounds. For the two N-compounds that did provide adhesion to dentin, NPG and NMNPG, the average bond strengths rose, peaked, and fell as the amounts of applied N-compound were increased. The molar efficiency dropped off as the concentration of applied N-compound rose. The least operator-sensitive and most efficient N-compound, NMNPG, delivered a bond strength equivalent to that of NPG, with 10% of the applied NPG amount.
Sigma-Aldrich
N-Phenylglycine, 97%