MilliporeSigma
  • Blockade of NMDA receptors in postnatal period decreased density of tyrosine hydroxylase immunoreactive axonal arbors in the medial prefrontal cortex of adult rats.
Blockade of NMDA receptors in postnatal period decreased density of tyrosine hydroxylase immunoreactive axonal arbors in the medial prefrontal cortex of adult rats.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2005-06-30)
K Wedzony, K Fijał, A Chocyk
ABSTRACT
Malfunction of glutamatergic neurotransmission in postnatal period is considered to be a risk factor for development of schizophrenia. Thus, the present study investigates the impact of NMDA receptor blockade in the postnatal period on the density of tyrosine hydroxylase immunoreactive axonal arbors in the rat medial prefrontal cortex. Behavioral experiments revealed that adult rats (60 days old) treated in the postnatal period with a competitive antagonist of NMDA receptors, CGP 40116 (1.25 mg/kg on days 1, 3, 6, 9; 2.5 mg/kg on days 12, 15, 18; and finally 5 mg/kg on day 21, all injections s.c.), showed enhancement of the locomotor activity stimulated by quinpirole (0.3 mg/kg s.c.) and amphetamine (0.5 mg/kg s.c.), which suggests development of functional supersensitivity of dopaminergic systems. It has been found that CGP 40116, given in postnatal period decreased the density of tyrosine hydroxylase immunoreactive axonal arbors in the medial prefrontal cortex of adult animals. The decrease was observed in superficial (II/III) and deep (V/VI) layers of the medial prefrontal cortex, while the average length of tyrosine hydroxylase immunoreactive axonal arbors was increased in both superficial and deep cortical layers. Changes in the density of tyrosine hydroxylase immunoreactive axonal arbors have not been followed by a significant decrease in the content of tyrosine hydroxylase protein measured by Western blot. Thus, NMDA receptor blockade in the early period of life evokes changes in architecture of tyrosine hydroxylase immunoreactive axonal arbors and that malfunction of glutamatergic neurotransmission, in early period of life may produce anatomical changes which resemble those observed in the brains of schizophrenics.
Sigma-Aldrich
CGP 37849, ≥98% (HPLC)