MilliporeSigma
Overtone spectra of aniline derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-12-13)
Vineet Kumar Rai, Amareswar Rai, D K Rai, S B Rai
ABSTRACT
Overtone spectra of 2-ethylaniline, N-methylaniline, N-ethylaniline, N,N-dimethylaniline and N,N-diethylaniline have been studied in 2500-15000 cm(-1) region. Vibrational frequency and anharmonicity constants for aryl/alkyl C-H stretch and N-H stretch vibrations have been determined. The effect of substitution of C(2)H(5) group on the ortho position in the ring and CH(3)/C(2)H(5) at the positions of the H-atom in NH(2) group has been studied in these molecules. It is noted that the aryl C-H stretching frequency and the N-H stretching frequency is appreciably increased due to the replacement of H in NH(2) group by CH(3)/C(2)H(5). These experimental observations are well supported by theoretical calculations for charge density on N-atom using molecular orbital AM(1) method.
Sigma-Aldrich
4-Ethylaniline, 98%