MilliporeSigma
  • Synthesis and structure-activity relationships of 1-arylmethyl-3-(1-methyl-2-amino)ethyl-5-aryl-6-methyluracils as antagonists of the human GnRH Receptor.
Synthesis and structure-activity relationships of 1-arylmethyl-3-(1-methyl-2-amino)ethyl-5-aryl-6-methyluracils as antagonists of the human GnRH Receptor.
Bioorganic & medicinal chemistry letters (2003-09-03)
Fabio C Tucci, Yun-Fei Zhu, Zhiqiang Guo, Timothy D Gross, Patrick J Connors, R Scott Struthers, Greg J Reinhart, John Saunders, Chen Chen
ABSTRACT
A new class of small molecule GnRH antagonists, the 1-arylmethyl-3-(1-methyl-2-amino)ethyl-5-aryl-6-methyluracils, was designed and a novel stereoselective synthesis for these compounds was developed. The stereochemical integrities of key intermediates (S)-6 and (R)-6 were confirmed by a combination of X-ray crystallography and chiral HPLC determinations. SAR studies were performed, which allowed the identification of derivatives (R)-9f, (R)-9h and (R)-12 as potent hGnRH antagonists (K(i)=20 nM).
Sigma-Aldrich
6-Methyluracil, 97%