MilliporeSigma
  • Development of a gas chromatographic method for fungicide cymoxanil analysis in dried hops.
Development of a gas chromatographic method for fungicide cymoxanil analysis in dried hops.
Journal of agricultural and food chemistry (2001-03-23)
M J Hengel, T Shibamoto
ABSTRACT
An analytical method for detecting cymoxanil [2-cyano-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide] residues in dried hops was developed utilizing liquid-liquid partitioning, automated gel permeation chromatography (GPC), solid phase extraction (SPE) cleanup, and gas chromatography (GC). Method validation recoveries from dried hops were 96 +/- 12, 108 +/- 11, and 136 +/- 8% over three levels of fortification (0.05, 0.5, and 1.0 ppm, respectively). The hop samples from three field sites, which were treated with cymoxanil, had residue levels ranging from 0.146 to 0.646 ppm. The detection limit and the quantitation limit of the method developed in the present study were 0.022 and 0.050 ppm, respectively.
Product Description
Supelco
Cymoxanil, PESTANAL®, analytical standard