MilliporeSigma
  • Clinical proof-of-concept study with MSDC-0160, a prototype mTOT-modulating insulin sensitizer.
Clinical proof-of-concept study with MSDC-0160, a prototype mTOT-modulating insulin sensitizer.
Clinical pharmacology and therapeutics (2013-03-07)
J R Colca, J T VanderLugt, W J Adams, A Shashlo, W G McDonald, J Liang, R Zhou, D G Orloff
ABSTRACT
It may be possible to achieve insulin sensitivity through the recently identified mitochondrial target of thiazolidinediones (mTOT), thereby avoiding peroxisome proliferator-activated receptor-γ (PPAR-γ)-dependent side effects. In this phase IIb clinical trial, 258 patients with type 2 diabetes completed a 12-week protocol with 50, 100, or 150 mg of MSDC-0160 (an mTOT modulator), 45 mg pioglitazone HCl (a PPAR-γ agonist), or a placebo. The two active treatments lowered fasting glucose levels to the same extent. The decreases in glycated hemoglobin (HbA1c) observed with the two higher doses of MSDC-0160 were not different from those associated with pioglitazone. By contrast, fluid retention as evidenced by reduction in hematocrit, red blood cells, and total hemoglobin was 50% less in the MSDC-0160-treated groups. There was also a smaller increase in high-molecular-weight (HMW) adiponectin with MSDC-0160 than with pioglitazone (P < 0.0001), suggesting that MSDC-0160 produces less expansion of white adipose tissue. Thus, mTOT modulators may have glucose-lowering effects similar to those of pioglitazone but without the adverse effects associated with PPAR-γ agonists.
Sigma-Aldrich
Pioglitazone hydrochloride, ≥98% (HPLC)