MilliporeSigma
  • Extraction of alumina and sodium oxide from red mud by a mild hydro-chemical process.
Extraction of alumina and sodium oxide from red mud by a mild hydro-chemical process.
Journal of hazardous materials (2009-09-09)
Li Zhong, Yifei Zhang, Yi Zhang
ABSTRACT
A mild hydro-chemical process to extract Al(2)O(3) in red mud to produce sodium aluminate hydrate was investigated, and the optimum conditions of Al(2)O(3) extraction were verified by experiments as leaching in 45% NaOH solution with CaO-to-red mud mass ratio of 0.25 and liquid-to-solid ratio of 0.9, under 0.8 MPa at 200 degrees C for 3.5h. Subsequent process of extracting Na(2)O from the residue of Al(2)O(3) extraction was carried out in 7% NaOH solution with liquid-to-solid ratio of 3.8 under 0.9 MPa at 170 degrees C for 2h. Overall, 87.8% of Al(2)O(3) and 96.4% of Na(2)O were extracted from red mud. The final residues with less than 1% Na(2)O could be utilized as feedstock in construction materials. The chemical reactions taking place in both Al(2)O(3) and Na(2)O extractions from red mud are proposed.
Sigma-Aldrich
Sodium oxide, 80%