MilliporeSigma
  • The beneficial effect of L-thyroxine on cardiovascular risk factors, endothelial function, and quality of life in subclinical hypothyroidism: randomized, crossover trial.
The beneficial effect of L-thyroxine on cardiovascular risk factors, endothelial function, and quality of life in subclinical hypothyroidism: randomized, crossover trial.
The Journal of clinical endocrinology and metabolism (2007-02-15)
Salman Razvi, Lorna Ingoe, Gill Keeka, Crispian Oates, Carolyn McMillan, Jolanta U Weaver
ABSTRACT
Subclinical hypothyroidism (SCH) is defined as raised serum TSH levels with circulating thyroid hormones within the reference range. It is uncertain whether treatment of SCH with L-thyroxine improves cardiovascular (CV) risk factors and quality of life. The objective of the study was to assess CV risk factors and patient-reported outcomes after treatment. This was a randomized, double-blind, crossover study of L-thyroxine and placebo. The study was conducted with community-dwelling patients. One hundred patients [mean age (sd) 53.8 (12) yr, 81 females] with SCH [mean TSH 6.6 (1.3) mIU/liter] without previously treated thyroid or vascular disease. Intervention consisted of 100 microg L-thyroxine or placebo daily for 12 wk each. Primary parameters were total cholesterol (TC) and endothelial function [brachial artery flow-mediated dilatation (FMD)], an early marker of atherosclerosis. Patient-reported outcomes were also assessed. L-thyroxine treatment reduced TC (vs. placebo) from 231.6 to 220 mg/dl, P < 0.001; low-density lipoprotein cholesterol from 142.9 to 131.3 mg/dl, P < 0.05; waist to hip ratio from 0.83 to 0.81, P < 0.006; and improved FMD from 4.2 to 5.9%, P < 0.001. Multivariate analysis showed that increased serum free T(4) level was the most significant variable predicting reduction in TC or improvement in FMD. Furthermore, the symptom of tiredness improved on L-thyroxine therapy, but other patient-reported outcomes were not significantly different after correction for multiple comparisons. SCH treated by L-thyroxine leads to a significant improvement in CV risk factors and symptoms of tiredness. The CV risk factor reduction is related to the increased level of achieved free T(4) concentration.