MilliporeSigma
  • [The combined treatment of patients with chronic nonspecific salpingo-oophoritis using a low-frequency magnetic field and iodobromine water].
[The combined treatment of patients with chronic nonspecific salpingo-oophoritis using a low-frequency magnetic field and iodobromine water].
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury (1998-12-18)
L P Markina, O V Iarustovskaia, L S Alisultanova, N A Derevnina, E V Gontar'
ABSTRACT
The studies of 116 females with chronic nonspecific salpingo-oophoritis demonstrate that combination of low-frequency magnetic field with iodine-bromine water effectively corrects immunological indices, i.e. improves T- and B-cell immunity, production of IgM and IgA, proportion of immunoregulatory subpopulations of T-lymphocytes.
Sigma-Aldrich
Iodine monobromide, 98%