MilliporeSigma
Reversible palmitoylation of the protein-tyrosine kinase p56lck.
The Journal of biological chemistry (1993-04-25)
L A Paige, M J Nadler, M L Harrison, J M Cassady, R L Geahlen
ABSTRACT
The myristoylated protein-tyrosine kinase, p56lck, is expressed predominantly in T cells where it is believed to play a role in T cell activation. We observed a 56-kDa protein that became metabolically labeled in intact T lymphoid cells that were incubated with either [3H]myristate or [3H]palmitate. This protein was identified as p56lck based on its specific immunoprecipitation with polyclonal antisera to p56lck, by induction of a shift in its electrophoretic mobility following treatment of cells with 12-O-tetradecanoylphorbol-13-acetate and by co-chromatography with p56lck on protamine-agarose. Characterization of the two acylation events revealed that, in contrast to the p56lck-associated radioactivity from [3H]myristate-labeled cells, the p56lck-associated radioactivity from [3H]palmitate-labeled cells was susceptible to cleavage by neutral hydroxylamine and was not blocked by inhibitors of protein synthesis. Pulse-chase analyses revealed that the labeling of p56lck with [3H]palmitate, but not [3H]myristate, was reversible. The presence of covalently attached palmitate on p56lck from [3H]palmitate-labeled cells was verified by thin-layer chromatography following acid hydrolysis of the acylated protein. 2-Hydroxymyristate, which is metabolically activated to form a potent inhibitor of protein myristoylation, specifically inhibited the acylation of p56lck with [3H]myristate without affecting its labeling with [3H]palmitate. These studies indicate that p56lck is both a cotranslationally myristoylated and post-translationally palmitoylated protein.
Sigma-Aldrich
2-Hydroxytetradecanoic acid, ≥98% (capillary GC)