MilliporeSigma
Reactive metabolites from the bioactivation of toxic methylfurans.
Science (New York, N.Y.) (1984-05-25)
V Ravindranath, L T Burka, M R Boyd
ABSTRACT
An important mechanism of toxicity of furans involves the cytochrome P-450 monooxygenase-catalyzed bioactivation of the compound in situ directly within the target tissues to highly reactive electrophilic products. The unsaturated aldehydes acetylacrolein and methylbutenedial have been identified as the principal reactive intermediates of 2- and 3-methylfuran, respectively, that are produced and bound covalently to tissue macromolecules in hepatic and pulmonary microsomal systems in vitro.
Sigma-Aldrich
2-Methylfuran, contains 200-400 BHT as stabilizer, 99%