MilliporeSigma
Identification of faceseal leak sites on a half-mask respirator.
American Industrial Hygiene Association journal (1990-05-01)
R K Oestenstad, J L Perkins, V E Rose
ABSTRACT
A method has been developed to identify the location and shape of respirator faceseal leak sites by the deposition of a fluorescent tracer. An aerosol generation, conditioning, and exposure system to provide a test environment with stable aerosol concentration and size distribution of 4-methyl-7-diethylaminocoumarin was designed and tested. Faceseal leak sites on a respirator mounted on a mannequin and worn by human subjects were identified by deposition of the tracer aerosol and subsequent observation under long-wave ultraviolet lighting. Test parameters were identified for the optimal definition of leaks. Photographic techniques were developed to document the identified leak sites.
Sigma-Aldrich
7-Diethylamino-4-methylcoumarin, 99%