MilliporeSigma
  • Imaging changes in synaptic acetylcholine availability in living human subjects.
Imaging changes in synaptic acetylcholine availability in living human subjects.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2012-11-20)
Irina Esterlis, Jonas O Hannestad, Frederic Bois, R Andrew Sewell, Rachel F Tyndale, John P Seibyl, Marina R Picciotto, Marc Laruelle, Richard E Carson, Kelly P Cosgrove
ABSTRACT
In vivo estimation of β(2)-nicotinic acetylcholine receptor availability with molecular neuroimaging is complicated by competition between the endogenous neurotransmitter acetylcholine and the radioligand (123)I-3-[2(S)-2-azetidinylmethoxy]pyridine ((123)I-5-IA). We examined whether binding of (123)I-5-IA is sensitive to increases in extracellular levels of acetylcholine in humans, as suggested in nonhuman primates. Six healthy subjects (31 ± 4 y) participated in a (123)I-5-IA SPECT study. After baseline scans, physostigmine (1-1.5 mg) was administered intravenously over 60 min, and 9 additional scans were obtained. We observed a significant reduction in the total volume of distribution after physostigmine administration (29% ± 17% in the cortex, 19% ± 15% in the thalamus, 19% ± 15% in the striatum, and 36% ± 30% in the cerebellum; P < 0.05). This reduction reflected a combination of a region-specific 7%-16% decrease in tissue concentration of tracer and a 9% increase in plasma parent concentration. These data suggest that increases in acetylcholine compete with (123)I-5-IA for binding to β(2)-nicotinic acetylcholine receptor. Additional validation of this paradigm is warranted, but it may be used to interrogate changes in extracellular acetylcholine.