MilliporeSigma
Rational design of light-directed dynamic spheres.
Chemical communications (Cambridge, England) (2012-11-02)
Yumi Okui, Mina Han
ABSTRACT
We created light-directed dynamic spheres based on simple azobenzene monomers showing (i) a high yield of reversible trans↔cis photoisomerization and (ii) noticeable phase transition from crystalline to isotropic states under UV light irradiation at ambient temperature.
Sigma-Aldrich
Azobenzene, 98%