MilliporeSigma
  • Palladium-catalyzed tandem reaction of o-aminophenols, bromoalkynes and isocyanides to give 4-amine-benzo[b][1,4]oxazepines.
Palladium-catalyzed tandem reaction of o-aminophenols, bromoalkynes and isocyanides to give 4-amine-benzo[b][1,4]oxazepines.
Chemical communications (Cambridge, England) (2012-10-23)
Bifu Liu, Yibiao Li, Meizhou Yin, Wanqing Wu, Huanfeng Jiang
ABSTRACT
A robust route to 4-amine-benzo[b][1,4]oxazepines relying upon a palladium-catalyzed tandem reaction of o-aminophenols, bromoalkynes and isocyanides has been developed. This chemistry presumably proceeds through the migratory insertion of isocyanides into the vinyl-palladium intermediate as a key step.
Sigma-Aldrich
2-Aminophenol, 99%