MilliporeSigma
  • Yeast-mediated xanthone synthesis through oxidative intramolecular cyclization.
Yeast-mediated xanthone synthesis through oxidative intramolecular cyclization.
Organic letters (2012-09-19)
Yann Fromentin, Philippe Grellier, Jean Duplex Wansi, Marie-Christine Lallemand, Didier Buisson
ABSTRACT
Benzoylphloroglucinol derivatives are natural products showing diverse biological activities that could be modulated by structural modifications. For this purpose, we studied the biotransformation of guttiferone A and of maclurin using a combinatorial approach for screening active microorganism strains. We found a novel and unexpected yeast-catalyzed oxidation that has selectively given a new oxy-guttiferone A and norathyriol.
Sigma-Aldrich
Xanthone, 97%