MilliporeSigma
  • In vitro anti-influenza virus and anti-inflammatory activities of theaflavin derivatives.
In vitro anti-influenza virus and anti-inflammatory activities of theaflavin derivatives.
Antiviral research (2012-04-24)
Mian Zu, Fan Yang, Weiling Zhou, Ailin Liu, Guanhua Du, Lishu Zheng
ABSTRACT
The theaflavins fraction (TF80%, with a purity of 80%) and three theaflavin (TF) derivatives from black tea have been found to exhibit potent inhibitory effects against influenza virus in vitro. They were evaluated with a neuraminidase (NA) activity assay, a hemagglutination (HA) inhibition assay, a real-time quantitative PCR (qPCR) assay for gene expression of hemagglutinin (HA) and a cytopathic effect (CPE) reduction assay. The experimental results showed that they all exerted significant inhibitory effects on the NA of three different subtypes of influenza virus strains [A/PR/8/34(H1N1), A/Sydney/5/97(H3N2) and B/Jiangsu/10/2003] with 50% inhibitory concentration (IC(50)) values ranging from 9.27 to 36.55 μg/mL, and they also displayed an inhibitory effect on HA; these inhibitory effects might constitute two major mechanisms of their antiviral activity. Time-of-addition studies demonstrated that TF derivatives might have a direct effect on viral particle infectivity, which was consistent with the inhibitory effect on HA. Subsequently, the inhibitory effect of TF derivatives on the replication of the viral HA gene as assayed by qPCR and on the nuclear localization of the influenza virus vRNP further demonstrated that they may primarily act during the early stage of infection. Interestingly, besides the activity against functional viral proteins, TF derivatives also decreased the expression level of the inflammatory cytokine IL-6 during viral infection, expression of which may result in serious tissue injury and apoptosis. Our results indicated that TF derivatives are potential compounds with anti-influenza viral replication and anti-inflammatory properties. These findings will provide important information for new drug design and development for the treatment of influenza virus infection.
Product Description
Supelco
Theaflavin, analytical standard