MilliporeSigma
  • Antidotes to anthrax lethal factor intoxication. Part 3: Evaluation of core structures and further modifications to the C2-side chain.
Antidotes to anthrax lethal factor intoxication. Part 3: Evaluation of core structures and further modifications to the C2-side chain.
Bioorganic & medicinal chemistry letters (2012-02-22)
Guan-Sheng Jiao, Seongjin Kim, Mahtab Moayeri, Devorah Crown, April Thai, Lynne Cregar-Hernandez, Linda McKasson, Banumathi Sankaran, Axel Lehrer, Teri Wong, Lisa Johns, Stephen A Margosiak, Stephen H Leppla, Alan T Johnson
ABSTRACT
Four core structures capable of providing sub-nanomolar inhibitors of anthrax lethal factor (LF) were evaluated by comparing the potential for toxicity, physicochemical properties, in vitro ADME profiles, and relative efficacy in a rat lethal toxin (LT) model of LF intoxication. Poor efficacy in the rat LT model exhibited by the phenoxyacetic acid series (3) correlated with low rat microsome and plasma stability. Specific molecular interactions contributing to the high affinity of inhibitors with a secondary amine in the C2-side chain were revealed by X-ray crystallography.
Sigma-Aldrich
Phenoxyacetic acid, 98%
Supelco
Phenoxyacetic acid, PESTANAL®, analytical standard