MilliporeSigma
  • Reactive oxygen species from chloroplasts contribute to 3-acetyl-5-isopropyltetramic acid-induced leaf necrosis of Arabidopsis thaliana.
Reactive oxygen species from chloroplasts contribute to 3-acetyl-5-isopropyltetramic acid-induced leaf necrosis of Arabidopsis thaliana.
Plant physiology and biochemistry : PPB (2012-02-07)
Shiguo Chen, Chunyan Yin, Reto Jörg Strasser, Govindjee, Chunlong Yang, Sheng Qiang
ABSTRACT
3-Acetyl-5-isopropyltetramic acid (3-AIPTA), a derivate of tetramic acid, is responsible for brown leaf-spot disease in many plants and often kills seedlings of both mono- and dicotyledonous plants. To further elucidate the mode of action of 3-AIPTA, during 3-AIPTA-induced cell necrosis, a series of experiments were performed to assess the role of reactive oxygen species (ROS) in this process. When Arabidopsis thaliana leaves were incubated with 3-AIPTA, photosystem II (PSII) electron transport beyond Q(A) (the primary plastoquinone acceptor of PSII) and the reduction of the end acceptors at the PSI acceptor side were inhibited; this was followed by increase in charge recombination and electron leakage to O(2), resulting in chloroplast-derived oxidative burst. Furthermore, the main antioxidant enzymes such as superoxide dismutase (SOD), catalase (CAT) and ascorbate peroxidase (APX) lost their activity. Excess ROS molecules directly attacked a variety of cellular components and subsequently caused electrolyte leakage, lipid peroxidation and cell membrane disruption. Finally, this led to cell destruction and leaf tissue necrosis. Thus, 3-AIPTA-triggered leaf necrosis of Arabidopsis was found to be a result of direct oxidative injury from the chloroplast-originated ROS burst initiated by the inhibition of normal photosynthetic electron transport.
Product Description
Supelco
Tenuazonic acid, analytical standard
Sigma-Aldrich
Tenuazonic acid copper salt from Alternaria alternata