MilliporeSigma
  • Atropa belladonna hairy roots: orchestration of concurrent oxidation and reduction reactions for biotransformation of carbonyl compounds.
Atropa belladonna hairy roots: orchestration of concurrent oxidation and reduction reactions for biotransformation of carbonyl compounds.
Applied biochemistry and biotechnology (2012-01-17)
Vikas Srivastava, Arvind Singh Negi, P V Ajayakumar, Shamshad A Khan, Suchitra Banerjee
ABSTRACT
The biotransformation potential of a selected Atropa belladonna hairy root clone (AB-09) had been evaluated with regard to three different aromatic carbonyl compounds, i.e., 3,4,5-trimethoxybenzaldehyde (1), 3,4,5-trimethoxyacetophenone (2), and 3,4,5-trimethoxy benzoic acid (3). The results demonstrated for the first time the untapped potentials of the selected hairy root clone to perform simultaneous oxidation (34.49%) and reduction (32.68%) of 3,4,5-trimethoxy benzaldehyde (1) into 3,4,5-trimethoxy benzoic acid (3), and 3,4,5-trimethoxy benzyl alcohol (4), respectively, without any intermediate separation or addition of reagents. The same hairy root clone also demonstrated reduction (<5%) of a 3,4,5-trimethoxyacetophenone (2) into a secondary alcohol, i.e., 1-(3,4,5-trimethoxyphenyl) ethanol (5), while in the case of aromatic carboxylic acid substrate (3), no biotransformation could be obtained under the similar conditions. The current observations revealed oxidation and reduction of the formyl group of the aromatic ring, and only reduction of the carbonyl group of acetophenone through the specific hairy root clone. The concurrent oxidation and reduction reactions by the selected hairy root clone highlight the importance of this study, which, as per our observations, is the first of its kind relating the hairy root culture of A. belladonna.
Sigma-Aldrich
3′,4′,5′-Trimethoxyacetophenone, 98%
Sigma-Aldrich
3,4,5-Trimethoxybenzoic acid, ReagentPlus®, 99%