MilliporeSigma
  • Exposure to nitric oxide increases the nitrosyl-iron complexes content in sorghum embryonic axes.
Exposure to nitric oxide increases the nitrosyl-iron complexes content in sorghum embryonic axes.
Plant science : an international journal of experimental plant biology (2011-12-27)
Marcela Simontacchi, Agustina Buet, Lorenzo Lamattina, Susana Puntarulo
ABSTRACT
This work was aimed to investigate nitrosyl-Fe complexes formation by reaction of endogenous ligands and Fe, in sorghum embryonic axes exposed to NO-donors. Electron paramagnetic resonance (EPR) was employed to detect the presence of nitrosyl-Fe complexes in plant embryos, as well as changes in labile iron pool (LIP). Nitrosyl-Fe complexes formation was detected in sorghum embryonic axes homogenates incubated in vitro in the presence of 1 mM of NO donors: diethylenetriamine NONOate (DETA NONOate), S-nitrosoglutathione (GSNO) and sodium nitroprusside (SNP). In axes isolated from seeds incubated in vivo in the presence of 1 mM SNP for 24 h, the content of NO was increased by 2-fold, and the EPR spectrum from mononitrosyl-Fe complexes (MNIC) was observed with a concomitant increase in the fresh weight of sorghum axes. The simultaneous exposure to deferoxamine and the NO donor precluded the increase in fresh weight observed in the presence of excess NO. While total Fe content in the axes isolated from seeds exposed to 1mM SNP was not significantly affected as compared to control axes, the LIP was increased by over 2-fold.The data reported suggest a critical role for the generation of complexes between Fe and NO when cells faced a situation leading to a significant increase in NO content. Moreover, demonstrate the presence of MNICs as one of the important components of the LIP, which could actively participate in Fe cellular mobilization.