MilliporeSigma
  • Novel stereoselective synthesis and chromatographic evaluation of E-guggulsterone.
Novel stereoselective synthesis and chromatographic evaluation of E-guggulsterone.
Steroids (2011-12-14)
Antimo Gioiello, Roccaldo Sardella, Emiliano Rosatelli, Bahman M Sadeghpour, Benedetto Natalini, Roberto Pellicciari
ABSTRACT
A new stereoselective synthesis of E-guggulsterone is described starting from androsten-3,17-dione. Protection of the ring A enonic system, followed by regioselective Wittig reaction and C-16 oxidation, affords E-guggulsterone in good yields and high stereoselectivity, making this approach easily accessible and scalable. Moreover, an original normal-phase HPLC method enabling the fast quantitation of the guggulsterone isomeric purity, combined with the suitability for sampling procedures, is detailed. The relying upon the cellulose-based Chiralpak IB column and the chloroform as the "non-standard" component of the eluent mixture, allows to get profitably high chromatographic performances.
Sigma-Aldrich
(Z)-Guggulsterone, ≥89% (HPLC), powder
Sigma-Aldrich
(E)-Guggulsterone, ≥95% (HPLC), powder